| Product Code | Brand | Quantity | Packaging | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
|---|---|---|---|---|---|---|---|---|---|
|
10509400
Safety data sheet (SDS) |
Fisher Chemical
T/3601/15
|
1 L | Amber Glass Bottle |
£175.81
1 L |
|
||||
|
10755921
Safety data sheet (SDS) |
Fisher Chemical
T/3601/17
|
2.5 L | Amber Glass Bottle |
£275.13
2.50 L |
|
||||
|
10579580
Safety data sheet (SDS) |
Fisher Chemical
T/3601/25
|
25 L | Metal Can |
£823.75
25 L |
|
2,2,4-Trimethylpentane, Certified AR for Analysis, Fisher Chemical™
£175.81 - £823.75
Chemical Identifiers
| CAS | 540-84-1 |
|---|---|
| Molecular Formula | C8H18 |
| Molecular Weight (g/mol) | 114.232 |
| MDL Number | 8943 |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| PubChem CID | 10907 |
| ChEBI | CHEBI:62805 |
| IUPAC Name | 2,2,4-trimethylpentane |
| SMILES | CC(C)CC(C)(C)C |
Specification
| Melting Point | -107°C |
|---|---|
| CAS | 540-84-1 |
| Color | Colorless |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Molecular Formula | C8H18 |
| Vapor Pressure | 51mbar at 20°C |
| MDL Number | 8943 |
| Viscosity | 0.51 mPaS at 22°C |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Synonym | isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane |
| Boiling Point | 98°C |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| SMILES | CC(C)CC(C)(C)C |
| IUPAC Name | 2,2,4-trimethylpentane |
| Molecular Weight (g/mol) | 114.232 |
| PubChem CID | 10907 |
| ChEBI | CHEBI:62805 |
| Formula Weight | 114.23g/mol |
Product Identifier
- 2,2,4-Trimethylpentane
Signal Word
- Danger
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H304- May be fatal if swallowed and enters airways.
- H315- Causes skin irritation.
- H336- May cause drowsiness or dizziness.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331- Do NOT induce vomiting.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental Information
- MIXTURE LIST- Contain: Isooctane