Implen™ DiluCell™ 10 - Disposable Cuvette
Implen DiluCell™ 10 provides an automatic 1/10 dilution of higher concentrated samples for standard 1 cm cuvette spectrophotometers. Combined with low sample volume requirements and bubble free filling, the DiluCell™ allows for convenient spectrophotometric analysis from 340-1100 nm.
Additional Details:
Weight:
0.10000kg
Disclaimers:
For Research Use Only
Specification
Description | 1/10 Dilution |
---|---|
Disposable | Single-use |
Capacity (Metric) | 200 μL |
Material | Plastic |
Quantity | Pack of 96 |
Path Length | 1.0 mm |
Wavelength Range | 340 to 1100 nm |
For Use With (Equipment) | Spectrophotometer, OD600 DiluPhotometer |
For Use With (Application) | Protein assays, OD600 |
Diameter (Metric) | 12 x 12 x 45 mm |