Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Marca:  Fisherbrand™ FB0129110
Codice prodotto. 30928568
  • 93.93€ / pezzi 1000
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Rib construction permits stacking without binding
  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.
Specifica
Specifica
50 mL
Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
5 mL
Autoclavable
Polypropylene
Beccuccio
Becher
Single-use