Learn More
Mettler Toledo™ LE pH Electrode LE438-IP67 pH/Conductivity/DO Meter
IP67 waterproof, plastic pH electrode with integrated temperature sensor.
Brand: Mettler Toledo™ 30247153
238.99 EUR valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Description
- Low maintainence
- For field use and for samples with fluctuating temperatures
- Plug/Connection: BNC/Cinch
Order Info
Measures pH only.
Specifications
LE438 Sensor; IP67 Waterproof | |
Field use, samples with fluctuating temperatures | |
Integrated temperature sensor | |
FiveGO, FiveEasy, and other Mettler Toledo pH Meters |