Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood
Provides an exceptional barrier against organic and inorganic chemicals and biological agents. Ansell Microchem™ 4000 Coveralls with Hood is constructed of a unique multi-layer fabric known for its lightwieght, yet robust textile feel.
Marca:  Ansell™ GR40T-00111-06
605.38 EUR valido fino al 2025-12-31
Usa il codice promo "24380" per avere il tuo prezzo promozionale.
Codice prodotto. 15516444
  • 929.00€ / pezzi 10
Solo null disponibili
Spedizione stimata: 18-07-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Coverall with elasticated 2-piece hood and front entry double-zip system.
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Protective Coveralls
Multi-layer Barrier Laminate Fabric
Elastic
Double, Elastic
2-panel, Elastic
Taped, Welded
None
2X-Large
Green
Elastic
Hooded
Double Zipper Front Entry
Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Includes Hood