missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen carbonate, 99%

Codice prodotto. p-7023444
Click to view available options
Quantità:
10,000 g
2500 g
500 g
Dimensione della confezione:
10000g
2500g
500g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 144-55-8
Formula molecolare CHNaO3
Molecular Weight (g/mol) 84.01
Numero MDL MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Sinonimo sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifica

Materiale o nome chimico Sodium hydrogen carbonate
pH 8.1 to 8.4
Quantità 500 g
Beilstein 4153970
Indice di Merck 14,8583
Informazioni di solubilità Soluble in water and methanol. Insoluble in ethanol.
Peso formulazione 84.01
Percent Purity 99%
Odore Odorless
Indice di rifrazione 1.5
Densità 2.159
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato