Learn More
Memmert™ Software AtmoCONTROL for UF110plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Marca:  Memmert™ UF110plusFDAQ1
Codice prodotto. 17578975
  • 2625.00€ / Pezzo
Solo null disponibili
Spedizione stimata: 08-08-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifica
Specifica
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
UF110plus Oven
Software