Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack
BIOPHARMAGREENER_CHOICE
Codice prodotto. 10150033
Cambia vista
Click to view available options
Capacità (sistema metrico):
3.4 mL
8 mL
15 mL
Finitura collo:
13-415 mm
20-415 mm
Dimensione della confezione:
pezzi 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Finitura collo unitSize
10150033 8 mL 20-415 mm pezzi 2000
30117591 3.4 mL 13-415 mm pezzi 2000
10181183 15 mL 20-415 mm pezzi 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10150033 Fornitore Thermo Scientific™ N. del fornitore 3620069025
Please to purchase this item. Need a web account? Register with us today!
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Safely store and handle test kits and many small volume applications with these PPCO diagnostic bottles, for use in sample storage and shipping small volume reagents.
Safely store and handle test kits and many small volume applications with Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Features:
  • Nonsterile
  • Natural translucent PPCO provides excellent chemical resistance
  • Autoclavable
  • Leakproof† when assembled with closure (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).
TRUSTED_SUSTAINABILITY
Specifica
Capacità 0.25 oz., 8 mL
Capacità (sistema metrico) 8 mL
Capacità (sistema imperiale) 0.25 oz.
Colore Translucent
Diametro (sistema metrico) interno collo 14 mm
Materiale PPCO
N. per custodia 2000
N. per confezione 2000
Sterilità Non-sterile
Finitura collo 20-415 mm
Certificazione/Conformità Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Bocca Narrow
Altezza (sistema metrico) 45 mm
Product Type Sample Container
Larghezza (sistema imperiale) 0.98 in.
Altezza (sistema imperiale) 1.77 in.
Diametro (sistema metrico) 25 mm
Erogazione attraverso la chiusura No
Autoclavabile Autoclavable
Vedi altri risultati Mostra meno risultati
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.