Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Marca:  Thermo Scientific™ 904-262
145.99 EUR valido fino al 2025-12-31
Usa il codice promo "24288" per avere il tuo prezzo promozionale.
 vedi altre versioni di questo prodotto
Codice prodotto. 10729601
  • 200.00€ / pezzi 960
Spedizione stimata: 13-08-2025
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
  • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifica
Specifica
1000 μl
Non sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Low Retention, Extended Length
960 Tips
Certified Quality
Non-filtered
Trasparente
ISO 9001 and ISO 13485
In rack
Reload Insert, 96 tips/insert, 10 inserts/pack
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus