Learn More
Falcon™ Pipette Controller, Standard Version
Sfoglia Tutto Corning Prodotti
Click to view available options
:
Standard Version
Descrizione
- Conveniently positioned switches quickly change operating modes and speeds to handle different liquid volumes and viscosities
- Aspirating and dispensing speed is controlled through the finger triggers
- When fully charged the batteries enable up to 8 hours of continuous use
- The large LCD display clearly indicates battery status, pipetting mode, and speed
- Additional filters are available as standard accessories
- Filters, pipette holder, and nosepiece are autoclavable
- Ergonomic design provides exceptional comfort
- Quick access to the battery compartment
- Easy maintenance
Specifica
Specifica
Descrizione | Suggested Replacement for 357470 |
Modello | Standard Version |
Numero di canali | 1 |
Product Line | Falcon |
Tipo | Serological |
Volume (sistema metrico) | 0.5 to 100 mL |
Display | LCD |
Tipo di filtro | (2) 0.2 μm and (2) 0.45 μm Hydrophobic Polytetrafluoroethylene (PTFE) Filters |
Da utilizzare con (apparecchiatura) | Glass and Plastic Serological Pipets with Volume 0.5 to 100 mL |
Include | 2-Position Charging Stand, Universal Power Supply, (4) Filters, (3) Batteries |