Learn More
Fisherbrand™ White Polypropylene Caps
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Finitura collo:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Rivestimento interno chiusura:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantità:
100
144
288
370
500
1300
2100
Descrizione
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifica
Specifica
Colore | White |
Quantità | 144 |
Colore chiusura | Bianco |
Finitura collo | 70-400 |
Closure Included | No |
Product Type | Lined Cap |
Rivestimento interno chiusura | PTFE/Polyethylene Foam |
Materiale chiusura | Polypropylene |