Code:
66
Falcon™ Plates in Ready-Stack Packaging
Versatility of plate colors to suit various detection methods
Additional Details:
Weight:
6.57710kg
Includes:
Lids
Specification
Array | 4 x 6 |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 3.5 mL |
Color | Clear |
Culture Area | 2 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Cell Culture Plate |
Volume (Metric) Working | 0.8 to 1 mL |
Well Count | 24 |
Includes | Lid |
No. per Case | 60/Cs. |
No. per Pack | 10/Pk. |
Packaging | 6 inner packs of 10 |
Quantity | 60/Cs. |