368.60 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Use promo code "24157" to get your promotional price.
Description | 12 Inserts |
---|---|
Culture Area | 0.33 cm2 |
Material | Polycarbonate |
Volume (Metric) Well | 0.6 mL |
Pore Size | 3 μm |
Type | Pre-loaded Microporous Insert in Tissue Culture Plate |
For Use With (Application) | Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies |
Diameter (Metric) Outer | 6.5 mm |
No. per Pack | 4 plates; 12 inserts/plate |
Surface Coating | TC-Treated |
Color | Clear |
Surface Treatment | TC-Treated |
Sterility | Sterile |
No. of Wells | 24 |