Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack
Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.
Overview
120.67 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: 03
Additional Details:
Additional Details: Weight: 1.17000kg
Product Code. 10029040
Quantity Price
1 £ 181.0 / Pack of 960
In stock, ready to ship: 142
In stock, ready to ship: 515
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Description ART 200, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 200 μL
Volume (Metric) Dispensing 200μL
Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Model ART 200
Length (English) 2.32 in
Length (Metric) 5.88 cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Universal
For Use With (Equipment) Universal Fit
Product Line ART
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes