Thermo Scientific Acros Isopropyl acetate, 99%, pure
CAS: 108-21-4 | C5H10O2 | 102.13 g/mol
Overview
322.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 108-21-4 Weight: 21.80000kg Transport: UN number : 1220 Chem class : 3 Pack group : II
Quantity 25 L
Packaging Metal Drum
Disclaimers: RUO – Research Use Only
Product Code. 10038571
Quantity Price
1 £ 323.61 / 25 L
Estimated Shipment
24-06-2025
Add to basket
Description and Specification
Chemical Identifiers
CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C
Specification
Chemical Name or Material Isopropyl acetate
Name Note 99%
CAS 108-21-4
Melting Point -73.0°C
Color Colorless
Density 0.8720g/mL
Boiling Point 88.8°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal Drum
Molecular Formula C5H10O2
Linear Formula CH3CO2CH(CH3)2
Refractive Index 1.3760 to 1.3780
MDL Number MFCD00008877
Quantity 25 L
Beilstein 02, 130
Merck Index 15, 5251
Specific Gravity 0.872
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Synonym isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
Solubility Information Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILES CC(C)OC(=O)C
IUPAC Name propan-2-yl acetate
Molecular Weight (g/mol) 102.13
PubChem CID 7915
Viscosity 0.49 mPa.s (25°C)
Formula Weight 102.13
Percent Purity 99%
Grade Pure
Physical Form Liquid