130.04 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Array | 4 x 6 |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 3.5 mL |
Color | Clear |
Culture Area | 2 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Cell Culture Plate |
Volume (Metric) Working | 1.5 to 2.2 mL |
Well Count | 24 |
Includes | Lid |
No. per Case | 50/Cs. |
No. per Pack | 1/Pk. |
Packaging | Individually Wrapped |
Pore Size | 1 μm |
Quantity | 50/Cs. |