Lenz Laborglasintrumente™ Needle-Valve Stopcock for Desiccators
Side tubulation needle-valve stopcock for desiccators
Additional Details:
Additional Details: Weight: 0.02000kg
Product Code. 10054320
Quantity Price
1 £ 37.8 / Each
Estimated Shipment
08-07-2025
Specification
Diameter (Metric) Outer 8 mm
No. per Pack 1
Description Cone: NS 24/29, Bore: NW 2.5 mm
Material Borosilicate Glass 3.3, PTFE
  • Cone: NS 24/29
  • Bore: NW 2.5 mm
  • Spindle: PTFE
  • Hose connection: outer diameter 8 mm
  • Material: borosilicate glass 3.3
  • Packaging unit: 1 piece
Alternative Products