Buerkle™ HDPE Wide-Necked Boxes, Round
Wide-necked containers with very good shatter resistance for convenient filling. Particularly suited for storing or shipping powders and granules.
Additional Details:
Weight:
0.07850kg
Specification
| Capacity (Metric) | 1500 mL |
|---|---|
| Color | Transparent |
| Diameter (Metric) Inner Neck | 67 mm |
| For Use With (Application) | Foodstuff |
| Includes | Screw Lid |
| Material | HDPE |
| Quantity | 1 |
| Weight (Metric) | 0.078 kg |
| Clarity | Transparent |
| Closure Included | Yes |
| Mouth | Wide |
| Shape | Round |
| Height (Metric) | 173 mm |
| Product Type | Wide Mouth Bottle |
| Closure Material | Polypropylene |
| Diameter (Metric) | 114 mm |
| Closure Type | Screw Cap |