Use promo code "25339" to get your promotional price.
Thermo Scientific Acros 1-Methylimidazole, 99%, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 5 kg |
---|---|
Packaging | Plastic drum |
Chemical Identifiers
CAS | 616-47-7 |
---|---|
Molecular Formula | C4H6N2 |
Molecular Weight (g/mol) | 82.11 |
MDL Number | MFCD00005292 |
InChI Key | MCTWTZJPVLRJOU-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
PubChem CID | 1390 |
ChEBI | CHEBI:113454 |
IUPAC Name | 1-methylimidazole |
SMILES | CN1C=CN=C1 |
Specification
Chemical Name or Material | 1-Methylimidazole |
---|---|
Name Note | 99% |
CAS | 616-47-7 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -2.0°C |
Color | Colorless to Yellow |
Density | 1.0300g/mL |
Boiling Point | 198.0°C |
Flash Point | 92°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic drum |
Molecular Formula | C4H6N2 |
Refractive Index | 1.4955 to 1.4975 |
MDL Number | MFCD00005292 |
Quantity | 5 kg |
Beilstein | 23, 46 |
Fieser | 05,447 |
Specific Gravity | 1.03 |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
Solubility Information | Solubility in water: 145g/L (25°C). Other solubilities: soluble in organic solvents |
InChI Key | MCTWTZJPVLRJOU-UHFFFAOYSA-N |
SMILES | CN1C=CN=C1 |
IUPAC Name | 1-methylimidazole |
Molecular Weight (g/mol) | 82.11 |
PubChem CID | 1390 |
ChEBI | CHEBI:113454 |
Formula Weight | 82.12 |
Percent Purity | 99% |
Physical Form | Liquid |
Technical documents
[1-Methylimidazole]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 1, sequence=3, description=Serious eye damage/eye irritation Category 1}, {code=SKIN CORR. 1B, sequence=4, description=Skin corrosion/irritation Category 1 B}, {code=REPR. 2, sequence=5, description=Reproductive toxicity Category 2}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H311, sequence=3, description=Toxic in contact with skin.}, {code=H302, sequence=4, description=Harmful if swallowed.}, {code=H361, sequence=5, description=Suspected of damaging fertility or the unborn child.}]
[{code=P280, sequence=50, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=51, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=54, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- 1-Methylimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 2
Hazard Statement
- H318- Causes serious eye damage.
- H314- Causes severe skin burns and eye damage.
- H311- Toxic in contact with skin.
- H302- Harmful if swallowed.
- H361- Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.