Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Aniline, 99.5%, extra pure
Use promo code "25339" to get your promotional price.
Quantity | 250 mL |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
CAS | 62-53-3 |
---|---|
Molecular Formula | C6H7N |
Molecular Weight (g/mol) | 93.13 |
MDL Number | MFCD00007629 |
InChI Key | PAYRUJLWNCNPSJ-UHFFFAOYSA-N |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
PubChem CID | 6115 |
ChEBI | CHEBI:17296 |
IUPAC Name | aniline |
SMILES | NC1=CC=CC=C1 |
Specification
Chemical Name or Material | Aniline |
---|---|
CAS | 62-53-3 |
Melting Point | -6.2°C |
Color | Brown to Colorless |
Density | 1.0213g/mL |
Boiling Point | 181.0°C to 185.0°C |
Flash Point | 76°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.4% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C6H7N |
Linear Formula | C6H5NH2 |
Refractive Index | 1.5845 to 1.5865 |
MDL Number | MFCD00007629 |
Quantity | 250 mL |
Beilstein | 12,59 |
Fieser | 06,21; 17,16 |
Merck Index | 15,652 |
Specific Gravity | 1.0213 |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Synonym | benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam |
Solubility Information | Solubility in water: 36g/L (20°C). |
InChI Key | PAYRUJLWNCNPSJ-UHFFFAOYSA-N |
SMILES | NC1=CC=CC=C1 |
IUPAC Name | aniline |
Molecular Weight (g/mol) | 93.13 |
PubChem CID | 6115 |
ChEBI | CHEBI:17296 |
Viscosity | 4.4 mPa.s (20°C) |
Formula Weight | 93.13 |
Percent Purity | 99.5% |
Grade | Extra Pure |
Physical Form | Liquid |
Technical documents
[Aniline]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}, {code=SKIN SENS. 1, sequence=3, description=Skin sensitiser Category 1}, {code=MUTA. 2, sequence=4, description=Germ cell mutagenicity Category 2}, {code=CARC. 2, sequence=5, description=Carcinogenicity Category 2}, {code=STOT RE 1, sequence=6, description=Specific target organ toxicity Category 1}, {code=AQUATIC ACUTE 1, sequence=7, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 1, sequence=8, description=LONG-TERM AQUATIC HAZARD Chronic 1}]
[{code=H301, sequence=1, description=Toxic if swallowed.}, {code=H311, sequence=2, description=Toxic in contact with skin.}, {code=H331, sequence=3, description=Toxic if inhaled.}, {code=H317, sequence=4, description=May cause an allergic skin reaction.}, {code=H318, sequence=5, description=Causes serious eye damage.}, {code=H341, sequence=6, description=Suspected of causing genetic defects.}, {code=H351, sequence=7, description=Suspected of causing cancer.}, {code=H372, sequence=8, description=Causes damage to organs.}, {code=H410, sequence=9, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P302+P352, sequence=51, description=IF ON SKIN: Wash with plenty of water/and soap.}, {code=P304+P340, sequence=52, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=54, description=Immediately call a POISON CENTER/doctor/.}, {code=P280, sequence=55, description=Wear protective gloves/protective clothing/eye protection/face protection.}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
- Aniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301- Toxic if swallowed.
- H311- Toxic in contact with skin.
- H331- Toxic if inhaled.
- H317- May cause an allergic skin reaction.
- H318- Causes serious eye damage.
- H341- Suspected of causing genetic defects.
- H351- Suspected of causing cancer.
- H372- Causes damage to organs.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P302+P352- IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.