Chemical Name or Material |
Sodium bicarbonate
|
Melting Point |
270.0°C
|
CAS |
144-55-8
|
Color |
White
|
Physical Form |
Powder
|
Quantity |
1 kg
|
Assay Percent Range |
99.7+%
|
Molecular Formula |
CHNaO3
|
Linear Formula |
NaHCO3
|
MDL Number |
MFCD00003528
|
Fieser |
03,260; 05,595; 11,476; 17,180
|
Merck Index |
15,8719
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
Solubility Information |
Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
|
InChI Key |
UIIMBOGNXHQVGW-UHFFFAOYSA-M
|
SMILES |
[Na+].OC([O-])=O
|
IUPAC Name |
sodium hydrogen carbonate
|
Molecular Weight (g/mol) |
84.01
|
PubChem CID |
516892
|
ChEBI |
CHEBI:32139
|
Formula Weight |
84.01
|
Percent Purity |
≥99.7%
|
Grade |
ACS Reagent
|
Packaging |
Plastic bottle
|