225.21 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Array | 2 x 3 |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 2.9 mL (Well) |
Color | Clear |
Culture Area | 9.5 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Multi-well Cell Culture Plate |
Volume (Metric) Working | 2 to 3 mL |
Well Count | 6 |
Includes | Lid |
No. per Case | 100/Cs. |
No. per Pack | 5/Pk. |
Packaging | Bulk |
Quantity | 100/Cs. |
Diameter (Metric) | 3.5 cm |