Code:
66
Corning™ Costar™ Ultra-Low Attachment Microplates
Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
Additional Details:
Weight:
1.62000kg
Specification
Array | 2 x 3 |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 2.9 mL (Well) |
Color | Clear |
Culture Area | 9.5 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Type | Multi-well Cell Culture Plate |
Well Count | 6 |
For Use With (Application) | Maintaining cells in a suspended, unattached state |
Includes | Lid |
No. per Case | 24/Cs. |
No. per Pack | 1/Pk. |
Packaging | Individually Wrapped |
Quantity | 24/Cs. |
Diameter (Metric) | 3.48 cm |