Code:
66
Corning™ 96-Well Half-Area Plates
Saves on valuable reagents by reducing the amount of reagent required per well, while retaining the ability to be read in standard microplate readers. Corning™ 96-Well Half Area Microplates are an economical solution for all of your assay diagnostic needs and are available in different colors and treatments.
Additional Details:
Weight:
5.85134kg
Specification
Barcode | Alphanumeric |
---|---|
Format | Half-Area |
Lid | Without Lid |
Array | 8 x 12 |
Packaging | 4 inner packs of 25 |
Color | Black |
For Use With (Equipment) | Plate readers |
Material | Polystyrene |
Volume (Metric) Well | 190 μL |
No. of Wells | 96 |
Volume (Metric) Working | 25 to 125 μL |
Sterility | Non-sterile |
Surface Treatment | Non-binding |
Well Shape | Flat Bottom (F-well) |
For Use With (Application) | Minimization of protein and nucleic acid binding |
No. per Case | 100/Cs. |
No. per Pack | 25/Pk. |
Quantity | 100/Cs. |
Well Color | Black |
-
Sample Preparation, Filtration and Storage
Get up to 80% off some of our most popular products.
Buy Now