199.64 GBP valid until 2025-12-31
Use promo code "24330" to get your promotional price.
Use promo code "24330" to get your promotional price.
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
Lid | With Lid |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Volume (Metric) Working | 50 to 250 μL/well |
Sterility | Sterile |
Surface Treatment | Nunclon Delta |
Well Shape | Flat Bottom (F-well) |
No. per Case | 50 |
No. per Pack | 1 |
Quantity | 50/Cs. |