Use promo code "24112" to get your promotional price.
Thermo Scientific™ PCR Plate, 96-well, low profile, non-skirted, white
Low profile 96-well plates for use in PCR and qPCR applications.
104.67 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Code:
03
Additional Details:
Weight:
1.00000kg
Specification
Barcode | No Barcode |
---|---|
Certifications/Compliance | Certified RNase/DNase-free |
Packaging | 25 Plate |
Color | White |
Product Type | PCR Microplate |
For Use With (Equipment) | Real-Time PCR Systems or Thermal Cyclers |
Skirt Style | Non-skirt |
Material | Polypropylene |
Volume (Metric) Well | 0.2 mL |
No. of Wells | 96 |
Volume (Metric) Working | 0.1 mL |
Quantity | 25 plates |
Reaction Speed | Low profile |