Use promo code "24140" to get your promotional price.
Fisher Chemical Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Use promo code "24140" to get your promotional price.
Quantity | 500 mL |
---|
Chemical Identifiers
CAS | 75-65-0 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | 4464 |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
IUPAC Name | 2-methylpropan-2-ol |
SMILES | CC(C)(C)O |
Specification
pH | 7 |
---|---|
Melting Point | 25°C |
CAS | 75-65-0 |
Boiling Point | 83°C |
Quantity | 500 mL |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Molecular Formula | C4H10O |
MDL Number | 4464 |
Vapor Pressure | 36mbar at 20°C |
Viscosity | 6.43 mPaS (25°C) |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
SMILES | CC(C)(C)O |
IUPAC Name | 2-methylpropan-2-ol |
Molecular Weight (g/mol) | 74.12 |
Formula Weight | 74.12g/mol |
Technical documents
[tert-Butanol]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H332, sequence=2, description=Harmful if inhaled.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H335, sequence=4, description=May cause respiratory irritation.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=55, description=Call a POISON CENTER/doctor/if you feel unwell.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- tert-Butanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H332- Harmful if inhaled.
- H319- Causes serious eye irritation.
- H335- May cause respiratory irritation.
- H336- May cause drowsiness or dizziness.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
-
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.
Buy Now