Use promo code "24112" to get your promotional price.
Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 10%, 1.0–1.5 mm
Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.
104.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Code:
50
Additional Details:
Weight:
0.90719kg
Includes:
One box of 10 gels
Specification
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
---|---|
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
Gel Percentage | 10% |
Gel Size | Mini |
GelThickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 25 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Wells | 10-well |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 gels (1 box) |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 8 cm |