438.90 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Barcode | No |
---|---|
Capacity (English) | 0.33 oz. |
Closure Color | Orange |
Closure Material | HDPE |
Capacity (Metric) | 10 mL |
Material | Polystyrene |
For Use With (Application) | For optimal cell attachment |
Includes | Lot numbers for ease in traceability |
Neck Style | Canted |
No. per Case | 200/cs. |
Shape | Rectangular |
Surface Coating | Corning Cellbind |
Graduated | Yes |
Sterility | Sterile |
Closure Type | Vented |