Use promo code "25339" to get your promotional price.
Fisher Chemical Ammonia Solution, 20%, Certified AR for Analysis, d=0.92, Fisher Chemical™
Use promo code "25339" to get your promotional price.
Quantity | 2.5 L |
---|
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
IUPAC Name | amine hydrate |
SMILES | N.O |
Specification
pH | 13.6 |
---|---|
Color | Colorless |
CAS | 1336-21-6 |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Quantity | 2.5 L |
SMILES | N.O |
IUPAC Name | amine hydrate |
Molecular Weight (g/mol) | 35.05 |
Packaging | Amber glass bottle |
Vapor Density | 0.59 |
Vapor Pressure | 500 hPa 50 |
Physical Form | Liquid |
Technical documents
[Ammonia solution 20/22%]
[Danger]
[{code=SKIN CORR. 1B, sequence=1, description=Skin corrosion/irritation Category 1 B}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H314, sequence=1, description=Causes severe skin burns and eye damage.}, {code=H335, sequence=2, description=May cause respiratory irritation.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ammonium hydroxide,Ammonia}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Ammonia solution 20/22%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314- Causes severe skin burns and eye damage.
- H335- May cause respiratory irritation.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : Ammonium hydroxide,Ammonia