94.24 GBP valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Use promo code "24108" to get your promotional price.
Content And Storage |
500 μL (50 applications of 10 μl each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red. Store at 4°C. |
---|---|
Detection Method | Colorimetric |
Number of Markers | 9 |
Product Type | Protein Ladder |
Ready to Load | Yes |
Size Range | 4 to 200 kDa |
Stain Type | 1 color: Blue |
Gel Compatibility | Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels |
Molecular Weight (g/mol) | 198, 62, 49, 38, 28, 18, 14, 6, 3 kDa |
Product Line | SeeBlue™ |
Quantity | 500 μL |
Shipping Condition | Wet Ice |
System Type | Western Blotting, SDS-PAGE |