Fisher Chemical Toluene, Extra Pure, SLR, Low In Sulfur, Fisher Chemical™
Overview
53.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Code: 21
Additional Details:
Additional Details: CAS Number: 108-88-3 Weight: 2.16500kg Transport: UN number : 1294 Chem class : 3 Pack group : II
Quantity 2.5 L
Packaging Amber Glass Bottle
Product Code. 10244020
Quantity Price
1 £ 84.8 / 2.50 L
In stock, ready to ship: 13
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
  • Special Offers
Description and Specification
Chemical Identifiers
CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1
Specification
Melting Point -95°C
CAS 108-88-3
Color Colorless
Boiling Point 111°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C7H8
MDL Number MFCD00008512
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
Synonym methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14g/mol
Alternative Products
Documentation Safety and Handling
[Toluene]
[Danger]
[{code=ASP. TOX. 1, sequence=1, description=Aspiration toxicity Category 1}, {code=FLAM. LIQ. 2, sequence=2, description=Flammable liquid Category 2}, {code=REPR. 2, sequence=3, description=Reproductive toxicity Category 2}, {code=SKIN IRRIT. 2, sequence=4, description=Skin corrosion/irritation Category 2}, {code=STOT RE 2, sequence=5, description=Specific target organ toxicity after repeated exposure Category 2}, {code=STOT SE 3, sequence=6, description=Specific target organ toxicity Category 3}]
[{code=H304, sequence=1, description=May be fatal if swallowed and enters airways.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H361d, sequence=3, description=Suspected of damaging the unborn child.}, {code=H373, sequence=4, description=May cause damage to organs.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}, {code=H225, sequence=6, description=Highly flammable liquid and vapour.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P310, sequence=52, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P331, sequence=55, description=Do NOT induce vomiting.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Toluene}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304- May be fatal if swallowed and enters airways.
  • H315- Causes skin irritation.
  • H361d- Suspected of damaging the unborn child.
  • H373- May cause damage to organs.
  • H336- May cause drowsiness or dizziness.
  • H225- Highly flammable liquid and vapour.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331- Do NOT induce vomiting.
Supplemental Information
  • MIXTURE LIST- Contains : Toluene
Special Offers