Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
For use with Branson ultrasonic baths
Additional Details:
Weight:
0.35000kg
Specification
Type | Perforated Tray |
---|---|
Capacity (Metric) | 11.4 L |
Capacity (English) | 2.5 gal. |
For Use With (Equipment) | Ultrasonic Bath, Model 5800 |
Material | Stainless Steel |
Dimensions (L x W x H) | 29.2 x 24.1 x 15.2 cm |