Use promo code "25339" to get your promotional price.
Fisher Chemical Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™
Use promo code "25339" to get your promotional price.
Quantity | 500 mL |
---|---|
Packaging | Amber glass bottle |
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | 8812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Specification
CAS | 1634-04-4 |
---|---|
Melting Point | -110°C |
Color | Colorless |
Boiling Point | 54°C |
Packaging | Amber glass bottle |
Molecular Formula | C5H12O |
MDL Number | 8812 |
Quantity | 500 mL |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
SMILES | CC(C)(C)OC |
IUPAC Name | 2-methoxy-2-methylpropane |
Molecular Weight (g/mol) | 88.15 |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
Formula Weight | 88.15g/mol |
Vapor Pressure | 268mbar at 20°C |
Physical Form | Liquid |
Technical documents
[tert-Butyl methyl ether]
[Danger]
[{code=SKIN IRRIT. 2, sequence=1, description=Skin corrosion/irritation Category 2}, {code=FLAM. LIQ. 2, sequence=2, description=Flammable liquid Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H315, sequence=2, description=Causes skin irritation.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=52, description=Ground/bond container and receiving equipment.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- tert-Butyl methyl ether
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H315- Causes skin irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.
- P302+P352- IF ON SKIN: Wash with plenty of water/.