92.15 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Barcode | No Barcode |
---|---|
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Format | Standard |
Lid | Without Lid |
Description | 96F well plate, Non-Treated, clear, without lid, Sterile (Pack of 1, Case of 50) |
Array | 8 x 12 |
Packaging | 1 Pk. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Skirt Style | Low Flange Pinchbar |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 50 |
No. per Pack | 1 |