Cytiva Ni Sepharose™ 6 Fast Flow
Combines the advantages of Ni2 for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform. Cytiva Lifescience™ Ni Sepharose™ 6 Fast Flow is part of a range of IMAC chromatography media for capture and intermediate purification.
Overview
Code: GE
Additional Details:
Additional Details: CAS Number: 7440-02-0 Weight: 0.06800kg
Product Code. 10291210
Quantity Price
1 £ 336.0 / 25mL
In stock, ready to ship: 261
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Buffer 0.1M HCl, 0.1M NaOH, 8M Urea
Product Type Ni Sepharose 6 FF chromatography media
Quantity 25 mL
Height (English) 9.84 in.
Height (Metric) 250mm
Flow Rate 600 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 12
Particle Size 90μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
dimMatrix Ni-Sepharose
Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.
  • Designed for scaling up purification of histidine-tagged proteins.
  • High protein binding capacity and minimal leakage of Ni2+ ions.
  • As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.
  • Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
  • The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Alternative Products
Safety and Handling
[Warning]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P210, sequence=52, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P241, sequence=53, description=Use explosion-proof electrical/ventilating/lighting/ equipment.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P235, sequence=55, description=Keep cool.}, {code=P501, sequence=56, description=Dispose of contents/container to .}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethanol, Nickel}]
[{code=GHS02, sequence=1, description=FLAME}]
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226- Flammable liquid and vapour.
Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241- Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235- Keep cool.
  • P501- Dispose of contents/container to .
Supplemental Information
  • MIXTURE LIST- Contains : Ethanol, Nickel