Fisher Chemical Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical
Anolyte solution for general use in conventional cells with a diaphragm
Overview
165.69 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 24
Additional Details:
Additional Details: CAS Number: 124-68-5 Weight: 0.87000kg Transport: UN number : 1992 Chem class : 3 Pack group : II
Product Code. 10302912
Quantity Price
1 £ 216.0 / 500mL
In stock, ready to ship: 693
Add to basket
Description and Specification
Specification
Chemical Name or Material Aqualine™ Electrolyte A
Color Colorless
Name Note Karl Fischer Reagent
Quantity 500 mL
CAS 67-66-3
Packaging Amber glass bottle
CAS 108-75-8
Physical Form Liquid
CAS 111-42-2
CAS 7446-09-5
Use Test Pass Test
MDL Number 81734
UN Number 1992
  • Anolyte solution for use in conventional cells with a diaphragm
  • Source of electrolyte required for anodic oxidation for coulometric analysis
  • Contains methanol and chloroform as solvents
Documentation Special Offers