Thermo Scientific™ Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Carry out daily laboratory work with Thermo Scientific™ Heratherm™ General Protocol Ovens, featuring temperature uniformity, easy settings and low energy consumption.
Overview
1703.87 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Code: 51
Additional Details:
Additional Details: Weight: 66.00000kg
Product Code. 10304150
Quantity Price
1 £ 2627.79 / Each
Estimated Shipment
24-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Accessories and Spare Parts
  • Special Offers
Description and Specification
Specification
Controller Type Microprocessor control with vacuum fluorescent display
Capacity (Metric) 176 L
Temperature (Metric) 50°C to 250°C
Type Gravity Convection
No. of Shelves 2 supplied/19 max.
Temperature Stability ±0.4°C
Timer ON / OFF
Dimensions (D x W x H) Interior 23.2 x 17.2 x 26.8 in. (58.9 x 43.8 x 68 cm)
Dimensions (L x W x H) 29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English) 29.1 in.
Length (Metric) 73.8 cm
Width (English) 25.2 in.
Width (English) Interior 17.2 in.
Width (Metric) 64 cm
Width (Metric) Interior 43.8 cm
Height (English) 36.2 in.
Height (English) Interior 26.8 in.
Height (Metric) 92 cm
Height (Metric) Interior 68 cm
Depth (English) Interior 23.2 in.
Depth (Metric) Interior 58.9 cm
Electrical Requirements 230 V 50/60 Hz
Voltage 230 V
Wattage 3100 w
Amperage 13.5 A
Frequency 50/60 Hz
Heratherm General Protocol Ovens provide a choice of gravity convection for gentle drying and heating, or mechanical convection for faster drying and heating, and superior temperature uniformity. Gravity convection units are designed to protect delicate samples while offering temperature uniformity of ±4.4°C or better, and temperature stability of ±0.4°C at 150°C. Mechanical convection models provide temperature uniformity of ±2.8°C or better, and temperature stability of at least ±0.3°C at 150°C.
  • Five sizes (60, 100 or 180, 400 or 750L)
  • Operating temperatures from 50° to 250°C
  • Automatic over-temperature alarm system to protect samples
  • Low overall energy consumption
  • Built in timer
  • Excellent footprint/volume ratio
  • Corrosion-resistant stainless-steel chambers with rounded corners for easy cleaning
  • Large, easy to read vacuum fluorescent display
  • Easy to use, microprocessor-controlled touch button operation
  • Easy to access—doors can be opened to over 180°C
Availability may vary by country.