Thermo Scientific Acros Aniline, 99.5%, extra pure
CAS: 62-53-3 | C6H7N | 93.13 g/mol
Overview
46.79 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 62-53-3 Weight: 2.55325kg Transport: UN number : 1547 Chem class : 6.1 Pack group : II
Quantity 2.5 L
Packaging Glass bottle
Disclaimers: RUO – Research Use Only
Product Code. 10318993
Quantity Price
1 £ 59.3 / 2.50 L
Estimated Shipment
22-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1
Specification
Chemical Name or Material Aniline
CAS 62-53-3
Melting Point -6.2°C
Color Brown to Colorless
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Molecular Formula C6H7N
Linear Formula C6H5NH2
Refractive Index 1.5845 to 1.5865
MDL Number MFCD00007629
Quantity 2.5 L
Beilstein 12,59
Fieser 06,21; 17,16
Merck Index 15,652
Specific Gravity 1.0213
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Synonym benzenamine,phenylamine,aminobenzene,aminophen,anilin,kyanol,arylamine,cyanol,benzeneamine,benzidam
Solubility Information Solubility in water: 36g/L (20°C).
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILES NC1=CC=CC=C1
IUPAC Name aniline
Molecular Weight (g/mol) 93.13
PubChem CID 6115
ChEBI CHEBI:17296
Viscosity 4.4 mPa.s (20°C)
Formula Weight 93.13
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Alternative Products
Documentation
Technical documents
Safety and Handling
[Aniline]
[Danger]
[{code=H301, sequence=1, description=Toxic if swallowed.}, {code=H311, sequence=2, description=Toxic in contact with skin.}, {code=H317, sequence=3, description=May cause an allergic skin reaction.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H331, sequence=5, description=Toxic if inhaled.}, {code=H341, sequence=6, description=Suspected of causing genetic defects.}, {code=H351, sequence=7, description=Suspected of causing cancer.}, {code=H372, sequence=8, description=Causes damage to organs.}, {code=H400, sequence=9, description=Very toxic to aquatic life.}, {code=H410, sequence=10, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Aniline}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
  • Aniline
Signal Word
  • Danger
Hazard Statement
  • H301- Toxic if swallowed.
  • H311- Toxic in contact with skin.
  • H317- May cause an allergic skin reaction.
  • H318- Causes serious eye damage.
  • H331- Toxic if inhaled.
  • H341- Suspected of causing genetic defects.
  • H351- Suspected of causing cancer.
  • H372- Causes damage to organs.
  • H400- Very toxic to aquatic life.
  • H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352- IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
  • MIXTURE LIST- Contains : Aniline