Use promo code "24140" to get your promotional price.
Ethylene glycol, 99+%, extra pure
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
32.64 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
Code:
29
Additional Details:
CAS Number:
107-21-1
Weight:
1.11300kg
| Quantity | 1 L |
|---|---|
| Packaging | Glass Bottle |
Disclaimers:
RUO – Research Use Only
Chemical Identifiers
| CAS | 107-21-1 |
|---|---|
| Molecular Formula | C2H6O2 |
| Molecular Weight (g/mol) | 62.068 |
| MDL Number | MFCD00002885 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| PubChem CID | 174 |
| ChEBI | CHEBI:30742 |
| IUPAC Name | ethane-1,2-diol |
| SMILES | C(CO)O |
Specification
| Chemical Name or Material | Ethylene glycol |
|---|---|
| Melting Point | -13°C |
| Name Note | 99+% |
| CAS | 107-21-1 |
| Decomposition Information | >500°C |
| Density | 1.1130g/mL |
| Quantity | 1 L |
| Flash Point | 116°C (240.8°F) |
| Infrared Spectrum | Authentic |
| Packaging | Glass Bottle |
| Physical Form | Viscous Liquid |
| Refractive Index | 1.4300 to 1.4330 (20°C, 589nm) |
| Assay Percent Range | 99+% |
| Molecular Formula | C2H6O2 |
| Linear Formula | HOCH2CH2OH |
| Vapor Pressure | Vapor Pressure: 0.12mmHg at 20°C |
| MDL Number | MFCD00002885 |
| Water | 0.1% max. (Coulometric) |
| Beilstein | 01, 465 |
| Fieser | 01,375; 05,296; 09,217; 15,156 |
| Merck Index | 15, 3852 |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Boiling Point | 196.0°C to 198.0°C |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| SMILES | C(CO)O |
| IUPAC Name | ethane-1,2-diol |
| Molecular Weight (g/mol) | 62.068 |
| PubChem CID | 174 |
| ChEBI | CHEBI:30742 |
| Formula Weight | 62.06 |
| Percent Purity | ≥99.0% |
| Grade | Extra Pure |
Technical documents
[Ethylene glycol]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=STOT RE 2, sequence=2, description=Specific target organ toxicity after repeated exposure Category 2}]
[{code=H373, sequence=1, description=May cause damage to organs.}, {code=H302, sequence=2, description=Harmful if swallowed.}]
[{code=P280, sequence=50, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P310, sequence=51, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Ethylene glycol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373- May cause damage to organs.
- H302- Harmful if swallowed.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /