CAS |
107-21-1
|
Melting Point |
-13°C
|
Color |
Colorless
|
pH |
5.5 to 7.5
|
Boiling Point |
196°C
|
Packaging |
HDPE plastic bottle
|
Molecular Formula |
C2H6O2
|
MDL Number |
2885
|
Quantity |
2.5 L
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
InChI Key |
LYCAIKOWRPUZTN-UHFFFAOYSA-N
|
SMILES |
C(CO)O
|
IUPAC Name |
ethane-1,2-diol
|
Molecular Weight (g/mol) |
62.068
|
PubChem CID |
174
|
Vapor Density |
2.14
|
ChEBI |
CHEBI:30742
|
Formula Weight |
62.06g/mol
|
Vapor Pressure |
0.12mmHg at 20°C
|
Physical Form |
Viscous Liquid
|