Macherey-Nagel™ MN 617 Grade Filter Papers, Folded
Suited for general filtration applications, including coarse-crystalline qualitative analysis. Macherey-Nagel™ MN 617 Grade Filter Papers, Folded are smooth, soft filter papers with fast filtration speed. The average ash content is about 0.1%, the amount of α-cellulose is about 95%.
Additional Details:
Weight:
0.10000kg
Specification
Basis Weight | 85 g/m2 |
---|---|
Description | Soft, smooth |
Format | Folded |
Material | Cellulose |
Particle Retention | ≥7 to 12μm |
Filtration Speed | 9sec. |
For Use With (Application) | Qualitative analysis and coarse-crystalline analysis |
Grade | MN 617 |
Quantity | 100 Pack |
Thickness (Metric) | 0.2 mm |
Type | Filter Paper |
Diameter (Metric) | 150 mm |