221.34 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Certifications/Compliance | ISO 9001:2008, ISO 13485:2003 |
---|---|
Capacity (Metric) | 75 cm² |
Material | Polystyrene,HDPE |
Material | Polystyrene,HDPE |
Neck Style | Angled |
No. per Case | 100 |
No. per Pack | 5 |
Product Type | Flask |
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Graduated | Yes |
Mouth Style | Narrow |
Sterility | Sterile |
Bottom Shape | Flat |
Closure Type | Filter |
Culture Area | 75 cm² |
Surface Treatment | Nunclon Delta |
Volume (Metric) Working | 25 mL |