Thermo Scientific™ TRACE™ TR-5 GC Columns
Choose these excellent starting columns for method development, capable of performing most required separations.
Overview
317.11 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 16
Additional Details:
Additional Details: Weight: 0.10000kg
Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10374603
Quantity Price
1 £ 335.0 / Each
Estimated Shipment
07-07-2025
Specification
Max. Temperature 320/340°C
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Product Line TRACE
Stationary Phase Trace™ TR-5
USP Type G27, G36
For Use With (Application) Aromatics Analysis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides
For Use With (Application) Aromatics Analysis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides
For Use With (Application) Aromatics Analysis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides
For Use With (Application) Aromatics Analysis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides
For Use With (Application) Aromatics Analysis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides
Film Thickness 0.25 μm
Length (Metric) 15 m
Polarity Non-Polar
Description 15 m x 0.25 mm x 0.25 μm
  • Nonpolar phase, 5% phenyl methyl polysiloxane
  • High operating temperature and extremely low-bleed
  • Widely used in a variety of applications
  • Similar to: DB-5, BP5, Rtx-5, HP-5, Ultra-2, PTE-5, SPB5, MDN-5, CP-Sil 8CB, SPB-5, AT-5, ZB-5, 007-2(MPS-5), SE-52, SE-54
Recommended for:
  • Alcohols
  • Free fatty acids
  • Aromatics
  • Flavors
  • Low polarity pesticides
Alternative Products