268.74 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Array | 4 x 6 |
---|---|
Closure Color | Clear |
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 0.57 mL (Well) |
Color | Clear |
Culture Area | 1.9 sq. cm |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Tissue Culture |
Type | Multi-well Cell Culture Plate |
Volume (Metric) Working | 0.5 mL |
Well Count | 24 |
Includes | Lid |
No. per Case | 100 (20 packs) |
No. per Pack | 5 Bulk Packed |
Packaging | Bulk |
Quantity | 100/Cs. |
Diameter (Metric) | 1.6 cm |