Use promo code "24127" to get your promotional price.
Fisher Chemical Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Use promo code "24127" to get your promotional price.
Quantity | 2.5 L |
---|---|
Packaging | Plastic Coated Glass bottle |
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Specification
pH | 1 |
---|---|
Color | Yellow |
CAS | 7697-37-2 |
Melting Point | -42°C |
Boiling Point | 122°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 2.5 L |
Molecular Formula | HNO3 |
Physical Form | Liquid |
MDL Number | MFCD00011349 |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
Synonym | hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
SMILES | O[N+]([O-])=O |
IUPAC Name | nitric acid |
Molecular Weight (g/mol) | 63.01 |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
Technical documents
[Nitric acid SG 1.42 (70%)]
[Danger]
[{code=OX. LIQ. 3, sequence=1, description=Oxidising liquid Category 3}, {code=MET. CORR. 1, sequence=2, description=Substances and mixtures corrosive to metals Category 1}, {code=SKIN CORR. 1A, sequence=3, description=Skin corrosion/irritation Category 1 A}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}]
[{code=H272, sequence=1, description=May intensify fire; oxidiser.}, {code=H290, sequence=2, description=May be corrosive to metals.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=EUH071, sequence=91, description=Corrosive to the respiratory tract.}]
[{code=P220, sequence=51, description=Keep/Store away from clothing/combustible materials.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=56, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=57, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Nitric acid}]
[{code=GHS03, sequence=1, description=FLAME OVER CIRCLE}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Nitric acid SG 1.42 (70%)
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272- May intensify fire; oxidiser.
- H290- May be corrosive to metals.
- H314- Causes severe skin burns and eye damage.
- EUH071- Corrosive to the respiratory tract.
Precautionary Statement
- P220- Keep/Store away from clothing/combustible materials.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : Nitric acid
-
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.
Buy Now