Code:
66
Corning™ Costar™ 24-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
Additional Details:
Weight:
3.62340kg
Specification
Array | 4 x 6 |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 0.57 mL (Well) |
Color | Clear |
Culture Area | 1.9 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Multi-well Cell Culture Plate |
Volume (Metric) Working | 0.5 mL |
Well Count | 24 |
Includes | Lid |
No. per Case | 50/Cs. |
No. per Pack | 1/Pk. |
Packaging | Individually Wrapped |
Quantity | 50/Cs. |
Diameter (Metric) | 1.6 cm |