Buerkle™ LDPE Wash Bottles with Swan-Neck Immersion Tube
Designed with severe curve in the neck to eliminate dangerous lateral spatter. Buerkle™ LDPE Wash Bottles with Swan-Neck Immersion Tube produces a fine, straight precision directed jet spray.
Overview
5.18 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.08100kg
Product Code. 10407311
Quantity Price
1 £ 5.45 / Each
In stock, ready to ship: 30
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Capacity (Metric) 1000 mL
Color Transparent
For Use With (Application) Used with Foodstuffs
Material LDPE
Height (Metric) 315 mm
Diameter (Metric) 98 mm
  • Manufactured from low density polyethylene (LDPE) for clarity, strength and flexibility
  • Immersion tube is removable
  • Translucent bottles for optimum visibility
  • Suitable for foodstuff
  • Screw cap with immersion tube