261.19 GBP valid until 2025-12-31
Use promo code "24322" to get your promotional price.
Use promo code "24322" to get your promotional price.
Capacity | 2 oz., 60 mL |
---|---|
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Color | Clear |
Description | Media bottles without closure |
Diameter (English) | 1.6 in. |
Diameter (English) Inner Neck | 0.71 in. |
Diameter (Metric) Inner Neck | 18 mm |
For Use With (Application) | Filling Lines |
Material | PETG |
No. per Case | 200 |
No. per Pack | 40 |
Sterility | Sterile |
Closure Included | No |
Graduated | Yes |
Mouth | Narrow |
Shape | Square |
Fit for Purpose | Sterile |
Height (Metric) | 41 mm |
Product Type | Media Bottle |
Height (English) | 1.6 in. |
Closure Size | 24-415 mm |
Diameter (Metric) | 41 mm |