Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Isopropyl acetate, 99%, pure
Use promo code "25339" to get your promotional price.
Quantity | 1 L |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
CAS | 108-21-4 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
MDL Number | MFCD00008877 |
InChI Key | JMMWKPVZQRWMSS-UHFFFAOYSA-N |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
PubChem CID | 7915 |
IUPAC Name | propan-2-yl acetate |
SMILES | CC(C)OC(=O)C |
Specification
Chemical Name or Material | Isopropyl acetate |
---|---|
Name Note | 99% |
CAS | 108-21-4 |
Melting Point | -73.0°C |
Color | Colorless |
Density | 0.8720g/mL |
Boiling Point | 88.8°C |
Flash Point | 4°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C5H10O2 |
Linear Formula | CH3CO2CH(CH3)2 |
Refractive Index | 1.3760 to 1.3780 |
MDL Number | MFCD00008877 |
Quantity | 1 L |
Beilstein | 02, 130 |
Merck Index | 15, 5251 |
Specific Gravity | 0.872 |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Synonym | isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester |
Solubility Information | Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents |
InChI Key | JMMWKPVZQRWMSS-UHFFFAOYSA-N |
SMILES | CC(C)OC(=O)C |
IUPAC Name | propan-2-yl acetate |
Molecular Weight (g/mol) | 102.13 |
PubChem CID | 7915 |
Viscosity | 0.49 mPa.s (25°C) |
Formula Weight | 102.13 |
Percent Purity | 99% |
Grade | Pure |
Physical Form | Liquid |
Technical documents
[Isopropyl acetate]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=EUH066, sequence=91, description=Repeated exposure may cause skin dryness or cracking.}]
[{code=P261, sequence=51, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P210, sequence=54, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=55, description=Ground/bond container and receiving equipment.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Isopropyl acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H319- Causes serious eye irritation.
- H336- May cause drowsiness or dizziness.
- EUH066- Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.